Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPLX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191350.1 | NP_001178279.1 | 1 - 2 | 160 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Bos taurus Probe spans exons |