Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191394.1 | NP_001178323.1 | 9 - 10 | 1799 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |