Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: N4BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191433.1 | NP_001178362.1 | 5 - 6 | 2291 | 91 | |
GenBank mRNA | EE896108.1 | - | 2 - 3 | 156 | 91 | |
GH319153.1 | - | 4 - 5 | 420 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |