Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191443.1 | NP_001178372.1 | 1 - 2 | 612 | 66 | |
GenBank mRNA | EE365078.1 | - | 1 - 2 | 119 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |