Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001245949.1 | NP_001232878.1 | 2 - 3 | 198 | 102 | |
GenBank mRNA | AB608099.1 | - | 2 - 3 | 436 | 102 | |
AB608100.1 | - | 2 - 3 | 198 | 102 | ||
LC271180.1 | - | 2 - 3 | 198 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Bos taurus Probe spans exons |