Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001245998.1 | NP_001232927.1 | 9 - 10 | 1378 | 79 | |
GenBank mRNA | GQ497280.2 | - | 9 - 10 | 1378 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Bos taurus Probe spans exons |