Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PABPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174568.3 | NP_776993.1 | 10 - 11 | 1959 | 64 | |
GenBank mRNA | AJ401269.2 | - | 10 - 11 | 64 | ||
BC102365.1 | - | 10 - 11 | 64 | |||
EE252871.1 | - | 3 - 4 | 444 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |