Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PABPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174568.3 | NP_776993.1 | 5 - 6 | 1257 | 101 | |
GenBank mRNA | AJ401269.2 | - | 5 - 6 | 743 | 101 | |
BC102365.1 | - | 5 - 6 | 1247 | 101 | ||
DV806286.1 | - | 3 - 4 | 265 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |