Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIDD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191487.1 | NP_001178416.1 | 1 - 2 | 308 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Bos taurus Probe spans exons |