Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191510.1 | NP_001178439.1 | 5 - 6 | 595 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |