Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRIT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191514.2 | NP_001178443.1 | 2 - 3 | 759 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Bos taurus Probe spans exons |