Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECT2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191539.1 | NP_001178468.1 | 5 - 6 | 769 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |