Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191541.1 | NP_001178470.1 | 7 - 8 | 1715 | 77 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Bos taurus Probe spans exons |