Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARRDC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192034.2 | NP_001178963.1 | 3 - 4 | 502 | 80 | |
GenBank mRNA | KP731623.1 | - | 3 - 4 | 628 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |