Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARRDC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192034.2 | NP_001178963.1 | 4 - 5 | 608 | 96 | |
GenBank mRNA | KP731623.1 | - | 4 - 5 | 734 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |