Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARRDC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192034.2 | NP_001178963.1 | 6 - 7 | 1028 | 67 | |
GenBank mRNA | KP731623.1 | - | 6 - 7 | 1154 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons |