Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR2H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192065.2 | NP_001178994.1 | 2 - 3 | 1348 | 109 | |
GenBank mRNA | BC118247.1 | - | 2 - 3 | 181 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |