Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNNI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192094.1 | NP_001179023.1 | 7 - 8 | 527 | 105 | |
GenBank mRNA | BC142168.1 | - | 7 - 8 | 507 | 105 | |
HQ697268.1 | - | 4 - 5 | 451 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |