Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192118.1 | NP_001179047.1 | 1 - 2 | 335 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Bos taurus Probe spans exons |