Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192129.1 | NP_001179058.1 | 1 - 2 | 143 | 83 | |
GenBank mRNA | BC113311.1 | - | 1 - 2 | 117 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |