Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192218.3 | NP_001179147.1 | 11 - 12 | 1595 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Bos taurus Probe spans exons |