Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192268.1 | NP_001179197.1 | 1 - 2 | 323 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Bos taurus Probe spans exons |