Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192278.2 | NP_001179207.2 | 5 - 6 | 1244 | 52 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |