Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PES1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001354818.1 | NP_001341747.1 | 9 - 10 | 996 | 73 | |
GenBank mRNA | BC118072.1 | - | 8 - 9 | 2337 | 73 | |
DV851730.1 | - | 1 - 2 | 91 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |