Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192321.1 | NP_001179250.1 | 12 - 13 | 1921 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |