Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFAP2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192329.1 | NP_001179258.1 | 4 - 5 | 1273 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |