Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFAP2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192329.1 | NP_001179258.1 | 7 - 8 | 1656 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Bos taurus Probe spans exons |