Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XKR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192331.1 | NP_001179260.1 | 2 - 3 | 945 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Bos taurus Probe spans exons |