Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TACR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192333.1 | NP_001179262.1 | 3 - 4 | 1447 | 107 | |
GenBank mRNA | AJ544580.1 | - | 3 - 4 | 495 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Bos taurus Probe spans exons |