Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192370.1 | NP_001179299.1 | 3 - 4 | 244 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |