Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192374.2 | NP_001179303.2 | 2 - 3 | 565 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |