Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EVX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192381.1 | NP_001179310.1 | 1 - 2 | 429 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Bos taurus Probe spans exons |