Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192393.3 | NP_001179322.1 | 2 - 3 | 811 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |