Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192405.2 | NP_001179334.1 | 2 - 3 | 352 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Bos taurus Probe spans exons |