Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPNE7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192413.1 | NP_001179342.1 | 6 - 7 | 734 | 70 | |
GenBank mRNA | DV883256.1 | - | 5 - 6 | 468 | 70 | |
EE344056.1 | - | 2 - 3 | 128 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |