Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM6SF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192430.1 | NP_001179359.1 | 2 - 3 | 305 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |