Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KBTBD12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192448.3 | NP_001179377.1 | 1 - 2 | 1068 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |