Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192452.1 | NP_001179381.1 | 2 - 3 | 583 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |