Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192892.1 | NP_001179821.1 | 7 - 8 | 1365 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |