Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192892.1 | NP_001179821.1 | 9 - 10 | 1657 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Bos taurus Probe spans exons |