Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GYS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192905.2 | NP_001179834.2 | 2 - 3 | 412 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons |