Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192934.3 | NP_001179863.1 | 2 - 3 | 941 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |