Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS26B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192958.2 | NP_001179887.1 | 5 - 6 | 1077 | 67 | |
GenBank mRNA | EV632220.1 | - | 2 - 3 | 207 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Bos taurus Probe spans exons |