Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192989.1 | NP_001179918.1 | 1 - 2 | 77 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |