Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMED8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193012.1 | NP_001179941.1 | 3 - 4 | 337 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons |