Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PALM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193034.3 | NP_001179963.2 | 1 - 2 | 94 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |