Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PALM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193034.3 | NP_001179963.2 | 3 - 4 | 2673 | 75 | |
NM_001371509.1 | NP_001358438.1 | 9 - 10 | 3385 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Bos taurus Probe spans exons |