Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193061.1 | NP_001179990.1 | 23 - 24 | 1828 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |