Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTDSPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193081.2 | NP_001180010.2 | 4 - 5 | 377 | 134 | |
GenBank mRNA | CK776230.1 | - | 4 - 5 | 377 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |