Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193121.1 | NP_001180050.1 | 1 - 2 | 47 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |